accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10294-29-8

IUPAC Name: gold(1+) chloride
Synonyms: gold chloride gold monochloride AuCl
Molecular Weight (g/mol): 232.42
Molecular Formula: AuCl
InChi Key: FDWREHZXQUYJFJ-UHFFFAOYSA-M
SMILES: [Cl-].[Au+]
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (4)
  • (4)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (7)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (4)
  • (4)
  • (7)
  • (3)