accessibility menu, dialog, popup

UserName

Viewing profile as user:
IAu

CAS RN 10294-31-2

IUPAC Name: gold(1+) iodide
Synonyms: gold monoiodide
Molecular Weight (g/mol): 323.87
Molecular Formula: AuI
InChi Key: ATGIETUGWDAYPU-UHFFFAOYSA-M
SMILES: [I-].[Au+]
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)