accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10294-50-5

IUPAC Name: tris(λ²-cobalt(2+)) octahydrate diphosphate
Synonyms: tris(λ²-cobalt(2+)) octahydrate diphosphate
Molecular Weight (g/mol): 510.86
Molecular Formula: Co3H16O16P2
InChi Key: QOQSIXJUYVUEMP-UHFFFAOYSA-H
SMILES: O.O.O.O.O.O.O.O.[Co++].[Co++].[Co++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results