accessibility menu, dialog, popup

UserName

Viewing profile as user:
KKOMnOOO

CAS RN 10294-64-1

IUPAC Name: dipotassium dioxomanganesebis(olate)
Synonyms: potassium manganate
Molecular Weight (g/mol): 197.13
Molecular Formula: K2MnO4
InChi Key: XKYFHUXZPRFUTH-UHFFFAOYSA-N
SMILES: [K+].[K+].[O-][Mn]([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)