accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1029439-85-7

IUPAC Name: 2,3-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Synonyms: 2,3-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Molecular Weight (g/mol): 256.06
Molecular Formula: C12H15BF2O3
InChi Key: JNBBZUYJDRTJCN-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C(F)C(F)=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results