accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1029612-18-7

CH3SOOOOOCH3CH3(E/Z)

CAS RN 1029612-18-7

IUPAC Name: methyl 3-[(4-methylbenzenesulfonyl)oxy]but-2-enoate
Synonyms: methyl 3-[(4-methylbenzenesulfonyl)oxy]but-2-enoate
Molecular Weight (g/mol): 270.3
Molecular Formula: C12H14O5S
InChi Key: QBENOPMVGLDWSB-UHFFFAOYSA-N
SMILES: COC(=O)C=C(C)OS(=O)(=O)C1=CC=C(C)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)