accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10297-05-9

IUPAC Name: 1-chloro-4-iodobutane
Synonyms: 1-chloro-4-iodobutane
Molecular Weight (g/mol): 218.46
Molecular Formula: C4H8ClI
InChi Key: JXOSPTBRSOYXGC-UHFFFAOYSA-N
SMILES: ClCCCCI
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)