accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaOOHN

CAS RN 103-01-5

IUPAC Name: sodium 2-(phenylamino)acetate
Synonyms: sodium DL-α-phenylaminoacetate
Molecular Weight (g/mol): 173.15
Molecular Formula: C8H8NNaO2
InChi Key: YEMGQZDWLLBIEY-UHFFFAOYSA-M
SMILES: [Na+].[O-]C(=O)CNC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (7)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (2)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (4)
  • (1)
  • (7)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)