accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 103-03-7

IUPAC Name: (phenylamino)urea
Synonyms: phenylaminourea
Molecular Weight (g/mol): 151.17
Molecular Formula: C7H9N3O
InChi Key: AVKHCKXGKPAGEI-UHFFFAOYSA-N
SMILES: NC(=O)NNC1=CC=CC=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)