CAS RN 103-05-9
IUPAC Name:
2-methyl-4-phenylbutan-2-ol
Synonyms:
benzenepropanol, α,α-dimethyl-
Molecular Weight (g/mol):
164.25
Molecular Formula:
C11H16O
InChi Key:
YXVSKJDFNJFXAJ-UHFFFAOYSA-N
SMILES:
CC(C)(O)CCC1=CC=CC=C1
1
–
1
of
1
results
1
2-Methyl-4-phenyl-2-butanol 99.0+%, TCI America™
CAS: 103-05-9 Molecular Formula: C11H16O Molecular Weight (g/mol): 164.25 MDL Number: MFCD00004472 InChI Key: YXVSKJDFNJFXAJ-UHFFFAOYSA-N Synonym: Dimethyl-beta-phenylethylcarbinol, 2-Phenylethyldimethylcarbinol PubChem CID: 7632 IUPAC Name: 2-methyl-4-phenylbutan-2-ol SMILES: CC(C)(O)CCC1=CC=CC=C1