accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 103-20-8

IUPAC Name: 7-ethyl-2-methylundecan-4-ol
Synonyms: 7-ethyl-2-methylundecan-4-ol
Molecular Weight (g/mol): 214.39
Molecular Formula: C14H30O
InChi Key: JKKBSZCOVNFRCQ-UHFFFAOYNA-N
SMILES: CCCCC(CC)CCC(O)CC(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results