accessibility menu, dialog, popup

UserName

Viewing profile as user:
N(E)

CAS RN 103-31-1

IUPAC Name: 4-[(1E)-2-phenylethenyl]pyridine
Synonyms: 4-[(1E)-2-phenylethenyl]pyridine
Molecular Weight (g/mol): 181.24
Molecular Formula: C13H11N
InChi Key: QKHRGPYNTXRMSL-VOTSOKGWSA-N
SMILES: C(=C/C1=CC=NC=C1)\C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results