accessibility menu, dialog, popup

UserName

Viewing profile as user:
NN(E/Z)

CAS RN 103-33-3

IUPAC Name: diphenyldiazene
Synonyms: diazobenzene
Molecular Weight (g/mol): 182.23
Molecular Formula: C12H10N2
InChi Key: DMLAVOWQYNRWNQ-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)N=NC1=CC=CC=C1
Boiling Point 
  • (6)
  • (2)
Percent Purity 
  • (3)
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (1)
  • (3)
  • (5)
  • (6)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (5)
  • (1)