accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OCH2

CAS RN 103-44-6

IUPAC Name: 1-(ethenyloxy)-2-ethylhexane
Synonyms: vinyl 2-ethylhexyl ether
Molecular Weight (g/mol): 156.27
Molecular Formula: C10H20O
InChi Key: DSSAWHFZNWVJEC-UHFFFAOYNA-N
SMILES: CCCCC(CC)COC=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results