accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOSOONH

CAS RN 103-47-9

IUPAC Name: sodium 2-(cyclohexylamino)ethane-1-sulfonate
Synonyms: ches (buffer)
Molecular Weight (g/mol): 229.27
Molecular Formula: C8H16NNaO3S
InChi Key: TXWKJOVDGFRVOG-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)CCNC1CCCCC1
Quantity 
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (1)
  • (3)
  • (2)
pH 
  • (1)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (8)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)