accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO(E)

CAS RN 103-53-7

IUPAC Name: 2-phenylethyl (2E)-3-phenylprop-2-enoate
Synonyms: phenethyl cinnamate
Molecular Weight (g/mol): 252.31
Molecular Formula: C17H16O2
InChi Key: MJQVZIANGRDJBT-VAWYXSNFSA-N
SMILES: O=C(OCCC1=CC=CC=C1)\C=C\C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results