accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 103-61-7

H3COO(E)

CAS RN 103-61-7

IUPAC Name: (2E)-3-phenylprop-2-en-1-yl butanoate
Synonyms: cinnamyl butyrate
Molecular Weight (g/mol): 204.27
Molecular Formula: C13H16O2
InChi Key: YZYPQKZWNXANRB-UXBLZVDNSA-N
SMILES: CCCC(=O)OC\C=C\C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results