accessibility menu, dialog, popup

UserName

Viewing profile as user:
Br(E)

CAS RN 103-64-0

IUPAC Name: [(1E)-2-bromoethenyl]benzene
Synonyms: β-bromostyrene
Molecular Weight (g/mol): 183.05
Molecular Formula: C8H7Br
InChi Key: YMOONIIMQBGTDU-VOTSOKGWSA-N
SMILES: Br\C=C\C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results