accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH

CAS RN 103-69-5

IUPAC Name: N-ethylaniline
Synonyms: ethylaniline N-ethylbenzeneamino N-ethylbenzenamino aethylanilin
Molecular Weight (g/mol): 121.18
Molecular Formula: C8H11N
InChi Key: OJGMBLNIHDZDGS-UHFFFAOYSA-N
SMILES: CCNC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)