accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3O

CAS RN 103-73-1

IUPAC Name: ethoxybenzene
Synonyms: phenyl ethyl alcohol
Molecular Weight (g/mol): 122.17
Molecular Formula: C8H10O
InChi Key: DLRJIFUOBPOJNS-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC=C1
Boiling Point 
  • (3)
  • (1)
  • (4)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (5)
  • (4)
Percent Purity 
  • (3)
  • (2)
  • (4)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (1)
  • (2)
  • (7)
  • (3)
  • (1)
  • (4)
  • (2)
  • (1)
  • (5)
  • (4)
  • (3)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (6)