accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHN

CAS RN 103-74-2

IUPAC Name: 2-(pyridin-2-yl)ethan-1-ol
Synonyms: 2-pyridineethanol
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: BXGYBSJAZFGIPX-UHFFFAOYSA-N
SMILES: OCCC1=CC=CC=N1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (5)