accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 103-80-0

IUPAC Name: 2-phenylacetyl chloride
Synonyms: phenylacetyl chloride
Molecular Weight (g/mol): 154.59
Molecular Formula: C8H7ClO
InChi Key: VMZCDNSFRSVYKQ-UHFFFAOYSA-N
SMILES: ClC(=O)CC1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (7)
Quantity 
  • (3)
  • (1)
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (2)
  • (4)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (7)
  • (3)
  • (1)
  • (1)
  • (4)
  • (2)