accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2SNH

CAS RN 103-85-5

IUPAC Name: phenylthiourea
Synonyms: N-phenylthiourea
Molecular Weight (g/mol): 152.22
Molecular Formula: C7H8N2S
InChi Key: FULZLIGZKMKICU-UHFFFAOYSA-N
SMILES: NC(=S)NC1=CC=CC=C1
Percent Purity 
  • (3)
  • (2)
  • (6)
Quantity 
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (5)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (1)
  • (2)
  • (6)
  • (3)
  • (2)
  • (6)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)