accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHBr

CAS RN 103-88-8

IUPAC Name: N-(4-bromophenyl)acetamide
Synonyms: 4-bromoacetanilide
Molecular Weight (g/mol): 214.06
Molecular Formula: C8H8BrNO
InChi Key: MSLICLMCQYQNPK-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(Br)C=C1
Percent Purity 
  • (2)
  • (7)
Quantity 
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (7)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)