accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 103-89-9

IUPAC Name: N-(4-methylphenyl)acetamide
Synonyms: 4-methylacetanilide
Molecular Weight (g/mol): 149.19
Molecular Formula: C9H11NO
InChi Key: YICAMJWHIUMFDI-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(C)C=C1
Boiling Point 
  • (3)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (2)
Quantity 
  • (3)
  • (2)
  • (3)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (3)
  • (3)
  • (2)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)