accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 103-95-7

IUPAC Name: 3-[4-(propan-2-yl)phenyl]butanal
Synonyms: cyclamen aldehyde
Molecular Weight (g/mol): 190.29
Molecular Formula: C13H18O
InChi Key: HQQPOVNESMNPNH-UHFFFAOYNA-N
SMILES: CC(C)C1=CC=C(C=C1)C(C)CC=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results