accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10300-69-3

IUPAC Name: hydrogen 2-chloroethanimidamide chloride
Synonyms: hydrogen 2-chloroethanimidamide chloride
Molecular Weight (g/mol): 128.98
Molecular Formula: C2H6Cl2N2
InChi Key: GUPOZVHRTJYZCX-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC(=N)CCl
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)