accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 103012-26-6

IUPAC Name: 3-phenyl-9H-carbazole
Synonyms: 3-phenyl-9H-carbazole
Molecular Weight (g/mol): 243.31
Molecular Formula: C18H13N
InChi Key: IAWRFMPNMXEJCK-UHFFFAOYSA-N
SMILES: N1C2=C(C=CC=C2)C2=C1C=CC(=C2)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results