accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSOONHN

CAS RN 10304-39-9

IUPAC Name: 4-methyl-N-(quinolin-8-yl)benzene-1-sulfonamide
Synonyms: 4-methyl-N-(quinolin-8-yl)benzenesulfonamide
Molecular Weight (g/mol): 298.36
Molecular Formula: C16H14N2O2S
InChi Key: ZSMKPYXVUIWTCT-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)NC1=C2N=CC=CC2=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results