accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 103044-68-4

IUPAC Name: 1-phenylnonadecan-1-one
Synonyms: 1-phenylnonadecan-1-one
Molecular Weight (g/mol): 358.61
Molecular Formula: C25H42O
InChi Key: WTWRNRBSHQDWMY-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCC(=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results