CAS RN 1030574-44-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-Chloro-2-[4-(dimethylamino)phenyl]benzimidazole, 95%, Thermo Scientific™
CAS: 1030574-44-7 MDL Number: MFCD15173053 Synonym: 4-(5-Chloro-2-benzimidazolyl)-N,N-dimethylaniline