accessibility menu, dialog, popup

UserName

Viewing profile as user:
SiBrBr

CAS RN 1030856-97-3

IUPAC Name: (3,5-dibromophenyl)triphenylsilane
Synonyms: (3,5-dibromophenyl)triphenylsilane
Molecular Weight (g/mol): 494.3
Molecular Formula: C24H18Br2Si
InChi Key: UYGFDYMFXPXXLL-UHFFFAOYSA-N
SMILES: BrC1=CC(=CC(Br)=C1)[Si](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results