accessibility menu, dialog, popup

UserName

Viewing profile as user:
PP

CAS RN 103099-52-1

IUPAC Name: dicyclohexyl[3-(dicyclohexylphosphanyl)propyl]phosphane
Synonyms: dicyclohexyl[3-(dicyclohexylphosphanyl)propyl]phosphane
Molecular Weight (g/mol): 436.65
Molecular Formula: C27H50P2
InChi Key: RSJBEKXKEUQLER-UHFFFAOYSA-N
SMILES: C(CP(C1CCCCC1)C1CCCCC1)CP(C1CCCCC1)C1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results