accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CNO

CAS RN 10315-98-7

IUPAC Name: 4-(2-methylpropyl)morpholine
Synonyms: N-isobutylmorpholine
Molecular Weight (g/mol): 143.23
Molecular Formula: C8H17NO
InChi Key: QKVSMSABRNCNRS-UHFFFAOYSA-N
SMILES: CC(C)CN1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results