accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClNH2SHOHO

CAS RN 10318-18-0

IUPAC Name: hydrogen 2-amino-3-sulfanylpropanoic acid chloride
Synonyms: hydrogen cysteine chloride
Molecular Weight (g/mol): 157.61
Molecular Formula: C3H8ClNO2S
InChi Key: IFQSXNOEEPCSLW-UHFFFAOYNA-N
SMILES: [H+].[Cl-].NC(CS)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)