accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1031929-01-7

IUPAC Name: 3-bromo-2-fluoro-5-(trifluoromethyl)pyridine
Synonyms: 3-bromo-2-fluoro-5-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 243.99
Molecular Formula: C6H2BrF4N
InChi Key: HKACHYLUXHBYMO-UHFFFAOYSA-N
SMILES: FC1=C(Br)C=C(C=N1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results