accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONONClN

CAS RN 10320-42-0

IUPAC Name: 2-chloro-5-nitropyrimidine
Synonyms: 2-chloro-5-nitropyrimidine
Molecular Weight (g/mol): 159.53
Molecular Formula: C4H2ClN3O2
InChi Key: OFCBNMYNAHUDGE-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CN=C(Cl)N=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results