accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10321-71-8

IUPAC Name: (2E)-4-methylpent-2-enoate
Synonyms: (2E)-4-methylpent-2-enoate
Molecular Weight (g/mol): 113.14
Molecular Formula: C6H9O2
InChi Key: QAOXMQCWUWZZNC-ONEGZZNKSA-M
SMILES: CC(C)\C=C\C([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)