accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3BrHO

CAS RN 10323-39-4

IUPAC Name: 4-bromo-2-tert-butylphenol
Synonyms: 4-bromo-2-tert-butylphenol
Molecular Weight (g/mol): 229.12
Molecular Formula: C10H13BrO
InChi Key: IKMJSWBFODAWTC-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(Br)=CC=C1O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results