accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1032358-02-3

IUPAC Name: 3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Synonyms: 3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol): 233.12
Molecular Formula: C13H20BNO2
InChi Key: PLCYEDJFSPVVLL-UHFFFAOYSA-N
SMILES: CC1=CN=CC(C)=C1B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results