CAS RN 10326-27-9
Barium Chloride Dihydrate (Crystalline/Certified ACS), Fisher Chemical™
CAS: 10326-27-9 Molecular Formula: BaCl2H4O2 Molecular Weight (g/mol): 244.26 MDL Number: MFCD00149154 InChI Key: PWHCIQQGOQTFAE-UHFFFAOYSA-L Synonym: barium chloride dihydrate,bacl2.2h2o,barium 2+ dihydrate dichloride,barium 2+ dichloride dihydrate,acmc-20alrc,dsstox_cid_131,barium chloride, acs,dsstox_rid_75389,dsstox_gsid_20131,ksc174q7d PubChem CID: 5284346 ChEBI: CHEBI:86153 IUPAC Name: barium(2+) dihydrate dichloride SMILES: O.O.[Cl-].[Cl-].[Ba++]
Barium Chloride TS/RS, 12% (w/v), Ricca Chemical
CAS: 10326-27-9 Molecular Formula: BaCl2H4O2 Molecular Weight (g/mol): 244.26 MDL Number: MFCD00149154 InChI Key: PWHCIQQGOQTFAE-UHFFFAOYSA-L Synonym: barium chloride dihydrate,bacl2.2h2o,barium 2+ dihydrate dichloride,barium 2+ dichloride dihydrate,acmc-20alrc,dsstox_cid_131,barium chloride, acs,dsstox_rid_75389,dsstox_gsid_20131,ksc174q7d PubChem CID: 5284346 ChEBI: CHEBI:86153 IUPAC Name: barium(2+) dihydrate dichloride SMILES: O.O.[Cl-].[Cl-].[Ba++]
Barium chloride dihydrate, ACS
CAS: 10326-27-9 Molecular Formula: BaCl2H4O2 Molecular Weight (g/mol): 244.26 MDL Number: MFCD00149154 InChI Key: PWHCIQQGOQTFAE-UHFFFAOYSA-L Synonym: barium chloride dihydrate,bacl2.2h2o,barium 2+ dihydrate dichloride,barium 2+ dichloride dihydrate,acmc-20alrc,dsstox_cid_131,barium chloride, acs,dsstox_rid_75389,dsstox_gsid_20131,ksc174q7d PubChem CID: 5284346 ChEBI: CHEBI:86153 SMILES: O.O.[Cl-].[Cl-].[Ba++]
Barium chloride dihydrate, 99+%
CAS: 10326-27-9 Molecular Formula: BaCl2H4O2 Molecular Weight (g/mol): 244.26 MDL Number: MFCD00149154 InChI Key: PWHCIQQGOQTFAE-UHFFFAOYSA-L Synonym: barium chloride dihydrate,bacl2.2h2o,barium 2+ dihydrate dichloride,barium 2+ dichloride dihydrate,acmc-20alrc,dsstox_cid_131,barium chloride, acs,dsstox_rid_75389,dsstox_gsid_20131,ksc174q7d PubChem CID: 5284346 ChEBI: CHEBI:86153 SMILES: O.O.[Cl-].[Cl-].[Ba++]
Barium chloride dihydrate, 99+%, ACS reagent
CAS: 10326-27-9 Molecular Formula: BaCl2H4O2 Molecular Weight (g/mol): 244.26 MDL Number: MFCD00149154 InChI Key: PWHCIQQGOQTFAE-UHFFFAOYSA-L Synonym: barium chloride dihydrate,bacl2.2h2o,barium 2+ dihydrate dichloride,barium 2+ dichloride dihydrate,acmc-20alrc,dsstox_cid_131,barium chloride, acs,dsstox_rid_75389,dsstox_gsid_20131,ksc174q7d PubChem CID: 5284346 ChEBI: CHEBI:86153 IUPAC Name: barium(2+) dihydrate dichloride SMILES: O.O.[Cl-].[Cl-].[Ba++]
Barium chloride dihydrate, Puratronic™, 99.997% (metals basis)
CAS: 10326-27-9 Molecular Formula: BaCl2H4O2 Molecular Weight (g/mol): 244.26 MDL Number: MFCD00149154 InChI Key: PWHCIQQGOQTFAE-UHFFFAOYSA-L Synonym: barium chloride dihydrate,bacl2.2h2o,barium 2+ dihydrate dichloride,barium 2+ dichloride dihydrate,acmc-20alrc,dsstox_cid_131,barium chloride, acs,dsstox_rid_75389,dsstox_gsid_20131,ksc174q7d PubChem CID: 5284346 ChEBI: CHEBI:86153 IUPAC Name: barium(2+) dihydrate dichloride SMILES: O.O.[Cl-].[Cl-].[Ba++]
Barium chloride, ≥99.0%, MP Biomedicals™
CAS: 10326-27-9 Molecular Formula: BaCl2H4O2 Molecular Weight (g/mol): 244.26 MDL Number: MFCD00149154 InChI Key: PWHCIQQGOQTFAE-UHFFFAOYSA-L Synonym: barium chloride dihydrate,bacl2.2h2o,barium 2+ dihydrate dichloride,barium 2+ dichloride dihydrate,acmc-20alrc,dsstox_cid_131,barium chloride, acs,dsstox_rid_75389,dsstox_gsid_20131,ksc174q7d PubChem CID: 5284346 ChEBI: CHEBI:86153 IUPAC Name: barium(2+) dihydrate dichloride SMILES: O.O.[Cl-].[Cl-].[Ba++]
Barium chloride dihydrate, 99+%, for analysis
CAS: 10326-27-9 Molecular Formula: BaCl2H4O2 Molecular Weight (g/mol): 244.26 MDL Number: MFCD00149154 InChI Key: PWHCIQQGOQTFAE-UHFFFAOYSA-L Synonym: barium chloride dihydrate,bacl2.2h2o,barium 2+ dihydrate dichloride,barium 2+ dichloride dihydrate,acmc-20alrc,dsstox_cid_131,barium chloride, acs,dsstox_rid_75389,dsstox_gsid_20131,ksc174q7d PubChem CID: 5284346 ChEBI: CHEBI:86153 IUPAC Name: barium(2+);dichloride;dihydrate SMILES: O.O.[Cl-].[Cl-].[Ba++]
Barium Chloride, Dihydrate, Crystal, ACS, 99%, Spectrum™ Chemical
CAS: 10326-27-9 Molecular Formula: BaCl2H4O2 Molecular Weight (g/mol): 244.26 MDL Number: MFCD00149154 InChI Key: PWHCIQQGOQTFAE-UHFFFAOYSA-L IUPAC Name: barium(2+) dihydrate dichloride SMILES: O.O.[Cl-].[Cl-].[Ba++]
Barium Chloride TS, (U.S.P. Test Solution), Spectrum™ Chemical
CAS: 10326-27-9 Molecular Formula: BaCl2H4O2 Molecular Weight (g/mol): 244.26 MDL Number: MFCD00149154 InChI Key: PWHCIQQGOQTFAE-UHFFFAOYSA-L IUPAC Name: barium(2+) dihydrate dichloride SMILES: O.O.[Cl-].[Cl-].[Ba++]
Barium Chloride, 10% (w/v), Ricca Chemical
CAS: 10326-27-9 Molecular Formula: BaCl2H4O2 Molecular Weight (g/mol): 244.26 MDL Number: MFCD00149154 InChI Key: PWHCIQQGOQTFAE-UHFFFAOYSA-L Synonym: barium chloride dihydrate,bacl2.2h2o,barium 2+ dihydrate dichloride,barium 2+ dichloride dihydrate,acmc-20alrc,dsstox_cid_131,barium chloride, acs,dsstox_rid_75389,dsstox_gsid_20131,ksc174q7d PubChem CID: 5284346 ChEBI: CHEBI:86153 IUPAC Name: barium(2+) dihydrate dichloride SMILES: O.O.[Cl-].[Cl-].[Ba++]
Barium Chloride, ACS Grade, ≥ 99.0%, LabChem™
CAS: 10326-27-9 Molecular Formula: BaCl2H4O2 Molecular Weight (g/mol): 244.26 MDL Number: MFCD00149154 InChI Key: PWHCIQQGOQTFAE-UHFFFAOYSA-L Synonym: barium chloride dihydrate,bacl2.2h2o,barium 2+ dihydrate dichloride,barium 2+ dichloride dihydrate,acmc-20alrc,dsstox_cid_131,barium chloride, acs,dsstox_rid_75389,dsstox_gsid_20131,ksc174q7d PubChem CID: 5284346 ChEBI: CHEBI:86153 IUPAC Name: barium(2+) dihydrate dichloride SMILES: O.O.[Cl-].[Cl-].[Ba++]
Barium Chloride, 2.00 N (1.00 M), Ricca Chemical
CAS: 10326-27-9 Molecular Formula: BaCl2H4O2 Molecular Weight (g/mol): 244.26 MDL Number: MFCD00149154 InChI Key: PWHCIQQGOQTFAE-UHFFFAOYSA-L Synonym: barium chloride dihydrate,bacl2.2h2o,barium 2+ dihydrate dichloride,barium 2+ dichloride dihydrate,acmc-20alrc,dsstox_cid_131,barium chloride, acs,dsstox_rid_75389,dsstox_gsid_20131,ksc174q7d PubChem CID: 5284346 ChEBI: CHEBI:86153 IUPAC Name: barium(2+) dihydrate dichloride SMILES: O.O.[Cl-].[Cl-].[Ba++]
Barium Chloride Solution R1 (61 g/L), Ricca Chemical
CAS: 10326-27-9 Molecular Formula: BaCl2H4O2 Molecular Weight (g/mol): 244.26 MDL Number: MFCD00149154 InChI Key: PWHCIQQGOQTFAE-UHFFFAOYSA-L Synonym: barium chloride dihydrate,bacl2.2h2o,barium 2+ dihydrate dichloride,barium 2+ dichloride dihydrate,acmc-20alrc,dsstox_cid_131,barium chloride, acs,dsstox_rid_75389,dsstox_gsid_20131,ksc174q7d PubChem CID: 5284346 ChEBI: CHEBI:86153 IUPAC Name: barium(2+) dihydrate dichloride SMILES: O.O.[Cl-].[Cl-].[Ba++]