accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10326-28-0

IUPAC Name: cadmium(2+) hexahydrate diperchlorate
Synonyms: cadmium(2+) hexahydrate diperchlorate
Molecular Weight (g/mol): 419.4
Molecular Formula: CdCl2H12O14
InChi Key: PQJZOTJHMRJOEM-UHFFFAOYSA-L
SMILES: O.O.O.O.O.O.[Cd++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results