accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3NH2Br

CAS RN 103273-01-4

IUPAC Name: 2-bromo-4-tert-butylaniline
Synonyms: 2-bromo-4-tert-butylaniline
Molecular Weight (g/mol): 228.13
Molecular Formula: C10H14BrN
InChi Key: OLKYFBNIFKQRIZ-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(Br)=C(N)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results