accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClOOONH

CAS RN 103321-49-9

IUPAC Name: (9H-fluoren-9-yl)methyl N-(2-chloro-2-oxoethyl)carbamate
Synonyms: 9H-fluoren-9-ylmethyl N-(2-chloro-2-oxoethyl)carbamate
Molecular Weight (g/mol): 315.75
Molecular Formula: C17H14ClNO3
InChi Key: HFWFBOCOXPEKBV-UHFFFAOYSA-N
SMILES: ClC(=O)CNC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)