accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10334-26-6

IUPAC Name: (1R,4S)-1,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione
Synonyms: (1R)-(-)-camphorquinone (1R)-bornane-2,3-dione
Molecular Weight (g/mol): 166.22
Molecular Formula: C10H14O2
InChi Key: VNQXSTWCDUXYEZ-LDWIPMOCSA-N
SMILES: CC1(C)[C@@H]2CC[C@@]1(C)C(=O)C2=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results