accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNOHNF

CAS RN 103342-82-1

IUPAC Name: 5-(2-fluorophenyl)-3-[(1H-indol-3-yl)methyl]-1-methyl-2,3-dihydro-1H-1,4-benzodiazepin-2-one
Synonyms: 5-(2-fluorophenyl)-3-(1H-indol-3-ylmethyl)-1-methyl-3H-1,4-benzodiazepin-2-one
Molecular Weight (g/mol): 397.45
Molecular Formula: C25H20FN3O
InChi Key: CGBANSGENFERAT-UHFFFAOYNA-N
SMILES: CN1C2=CC=CC=C2C(=NC(CC2=CNC3=CC=CC=C23)C1=O)C1=CC=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results