accessibility menu, dialog, popup

UserName

Viewing profile as user:
FSClOO

CAS RN 103360-04-9

IUPAC Name: (4-fluorophenyl)methanesulfonyl chloride
Synonyms: (4-fluorophenyl)methanesulfonyl chloride
Molecular Weight (g/mol): 208.63
Molecular Formula: C7H6ClFO2S
InChi Key: UUQGWVIRPCRTSA-UHFFFAOYSA-N
SMILES: FC1=CC=C(CS(Cl)(=O)=O)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results