accessibility menu, dialog, popup

UserName

Viewing profile as user:
ON

CAS RN 10338-57-5

IUPAC Name: 4-(piperidin-1-yl)benzaldehyde
Synonyms: 4-(piperidin-1-yl)benzaldehyde
Molecular Weight (g/mol): 189.26
Molecular Formula: C12H15NO
InChi Key: ILJVPSVCFVQUAD-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(C=C1)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results