accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1034-39-5

IUPAC Name: dibromotriphenyl-λ⁵-phosphane
Synonyms: triphenylphosphine dibromide
Molecular Weight (g/mol): 422.1
Molecular Formula: C18H15Br2P
InChi Key: OCXGTPDKNBIOTF-UHFFFAOYSA-N
SMILES: BrP(Br)(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)