accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNOSSOONa(E/Z)

CAS RN 103404-75-7

IUPAC Name: sodium 2-(6-oxo-2,6-dihydro-1,3-benzothiazol-2-ylidene)-1,3-thiazolidine-4-carboxylate
Synonyms: sodium 2-(6-oxo-1,3-benzothiazol-2-ylidene)-1,3-thiazolidine-4-carboxylate
Molecular Weight (g/mol): 302.3
Molecular Formula: C11H7N2NaO3S2
InChi Key: BZNVUYVALNTPBG-UHFFFAOYNA-M
SMILES: [Na+].[O-]C(=O)C1CSC(N1)=C1SC2=CC(=O)C=CC2=N1
Color 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)